siRNA Details
| Gene Symbol: Flnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081185.1 | NP_001074654.1 | 39, 40 | 6798 | ||
| GenBank mRNA | AK135523.1 | - | 4, 5 | 638 | ||
| BC039885.1 | - | 4, 5 | 666 | |||
| BC052186.1 | - | 12, 13 | 1747 | |||
| BC060276.1 | - | 18, 19 | 2859 | |||
| BC151097.1 | - | 39, 40 | 6603 | 9088010 | ||
| BC158128.1 | - | 38, 39 | 6504 | 9007396 | ||
| BC172159.1 | - | 38, 39 | 6504 | |||

Flnc
Genomic Map