siRNA Details
Gene Symbol: Foxk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080932.2 | NP_001074401.2 | 9 | 3366 | ||
GenBank mRNA | AK013315.1 | - | 1 | 485 | ||
AK017613.1 | - | 1 | 754 | |||
AK150439.1 | - | 8 | 2810 |
