siRNA Details
| Gene Symbol: Dtwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170960.1 | NP_001164431.1 | 4 | 2524 | ||
| NM_026854.3 | NP_081130.1 | 6 | 2812 | |||
| GenBank mRNA | AK033232.1 | - | 1 | 1058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtwd2| Gene Symbol: Dtwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170960.1 | NP_001164431.1 | 4 | 2524 | ||
| NM_026854.3 | NP_081130.1 | 6 | 2812 | |||
| GenBank mRNA | AK033232.1 | - | 1 | 1058 | ||