siRNA Details
| Gene Symbol: Dtwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170960.1 | NP_001164431.1 | 3 | 485 | ||
| NM_026854.3 | NP_081130.1 | 5 | 773 | |||
| GenBank mRNA | AK004450.1 | - | 5 | 748 | ||
| AK144689.1 | - | 5 | 748 | |||
| BC075693.1 | - | 7 | 867 | |||
| BC139347.1 | - | 5 | 739 | 8862369 | ||
| BC139348.1 | - | 5 | 739 | 8862370 | ||
| BC145608.1 | - | 4 | 546 | |||
| BC145609.1 | - | 3 | 451 | 9054175 | ||

Dtwd2
Genomic Map