siRNA Details
| Gene Symbol: Smim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163721.1 | NP_001157193.1 | 4 | 1781 | ||
| NM_001163722.1 | NP_001157194.1 | 4 | 1707 | |||
| GenBank mRNA | AK028394.1 | - | 5 | 1831 | ||
| AK038662.1 | - | 4 | 3327 | |||
| AK158109.1 | - | 2 | 3549 | |||
| AK165247.1 | - | 4 | 1679 | |||
| AK167398.1 | - | 4 | 1696 | |||
| BC055936.1 | - | 3 | 1563 | |||
| BC080682.1 | - | 2 | 1874 | |||

Smim1
Genomic Map