siRNA Details
| Gene Symbol: Smim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163721.1 | NP_001157193.1 | 4 | 2327 | ||
| NM_001163722.1 | NP_001157194.1 | 4 | 2253 | |||
| GenBank mRNA | AK028394.1 | - | 5 | 2377 | ||
| AK038662.1 | - | 4 | 3873 | |||
| AK158109.1 | - | 2 | 4095 | |||
| AK165247.1 | - | 4 | 2225 | |||
| AK167398.1 | - | 4 | 2242 | |||
| BC055936.1 | - | 3 | 2109 | |||
| BC080682.1 | - | 2 | 2420 | |||

Smim1
Genomic Map