siRNA Details
| Gene Symbol: Gkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026860.1 | NP_081136.1 | 1 | 81 | ||
| GenBank mRNA | BC116955.1 | - | 1 | 81 | 40126274 | |
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Gkn3
Gkn3| Gene Symbol: Gkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026860.1 | NP_081136.1 | 1 | 81 | ||
| GenBank mRNA | BC116955.1 | - | 1 | 81 | 40126274 | |
 Back To Top
 Back To Top