siRNA Details
| Gene Symbol: Gkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026860.1 | NP_081136.1 | 6 | 673 | ||
| GenBank mRNA | AK004474.1 | - | 6 | 593 | ||
| BC116955.1 | - | 6 | 673 | 40126274 | ||
| FN600638.1 | - | 6 | 592 | |||
| GU220566.1 | - | 6 | 592 | |||

Gkn3
Genomic Map