siRNA Details
| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026861.2 | NP_081137.2 | 5 | 625 | ||
| GenBank mRNA | AK035384.1 | - | 2 | 200 | ||
| AK049109.1 | - | 5 | 574 | |||
| AK158177.1 | - | 5 | 574 | |||
| AK199665.1 | - | 3 | 311 | |||
| BC024467.1 | - | 5 | 611 | 4975777 | ||

Ubac2
Genomic Map