siRNA Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019253.3 | NP_062126.3 | 4 | 969 | ||
| XM_006229252.2 | XP_006229314.1 | 9 | 1874 | |||
| XM_006229253.2 | XP_006229315.1 | 9 | 1815 | |||
| XM_006229254.2 | XP_006229316.1 | 8 | 1714 | |||
| XM_006229255.2 | XP_006229317.1 | 8 | 1790 | |||
| XM_006229257.2 | XP_006229319.1 | 7 | 1621 | |||
| XM_008759346.1 | XP_008757568.1 | 9 | 1349 | |||
| GenBank mRNA | BC092196.1 | - | 2 | 334 | 7313460 | |
| FQ211673.1 | - | 9 | 1478 | |||
| FQ213308.1 | - | 6 | 783 | |||
| S49400.1 | - | 4 | 969 | |||

Ptpn5
Genomic Map