siRNA Details
| Gene Symbol: Zfp707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081065.1 | NP_001074534.1 | 4 | 1002 | ||
| GenBank mRNA | AK137962.1 | - | 3 | 1885 | ||
| AK153958.1 | - | 4 | 952 | |||
| BC026404.1 | - | 4 | 968 | 4210825 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp707| Gene Symbol: Zfp707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081065.1 | NP_001074534.1 | 4 | 1002 | ||
| GenBank mRNA | AK137962.1 | - | 3 | 1885 | ||
| AK153958.1 | - | 4 | 952 | |||
| BC026404.1 | - | 4 | 968 | 4210825 | ||