siRNA Details
| Gene Symbol: Zfp707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081065.1 | NP_001074534.1 | 4 | 863 | ||
| GenBank mRNA | AK005330.1 | - | 4 | 812 | ||
| AK137962.1 | - | 3 | 1746 | |||
| AK153958.1 | - | 4 | 813 | |||
| BC026404.1 | - | 4 | 829 | 4210825 | ||

Zfp707
Genomic Map