siRNA Details
| Gene Symbol: 1810013L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081400.3 | NP_001074869.1 | 4 | 2713 | ||
| GenBank mRNA | AK032072.1 | - | 1 | 1122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810013L24Rik| Gene Symbol: 1810013L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081400.3 | NP_001074869.1 | 4 | 2713 | ||
| GenBank mRNA | AK032072.1 | - | 1 | 1122 | ||