siRNA Details
| Gene Symbol: 1810011H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163616.1 | NP_001157088.1 | 5 | 662 | ||
| GenBank mRNA | AK007434.1 | - | 5 | 663 | ||
| AK014636.1 | - | 4 | 603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810011H11Rik| Gene Symbol: 1810011H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163616.1 | NP_001157088.1 | 5 | 662 | ||
| GenBank mRNA | AK007434.1 | - | 5 | 663 | ||
| AK014636.1 | - | 4 | 603 | |||