siRNA Details
Gene Symbol: 1810011H11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163616.1 | NP_001157088.1 | 5 | 448 | ||
GenBank mRNA | AK007434.1 | - | 5 | 449 | ||
AK014636.1 | - | 4 | 389 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810011H11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163616.1 | NP_001157088.1 | 5 | 448 | ||
GenBank mRNA | AK007434.1 | - | 5 | 449 | ||
AK014636.1 | - | 4 | 389 |