siRNA Details
| Gene Symbol: 1810022K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099674.1 | NP_001093144.1 | 3 | 178 | ||
| GenBank mRNA | AK007585.1 | - | 3 | 179 | ||
| BC045157.1 | - | 3 | 165 | |||
| BC055766.1 | - | 3 | 166 | |||
| BC083104.1 | - | 3 | 177 | |||
| BC100487.1 | - | 3 | 174 | |||
| BC100488.1 | - | 3 | 162 | |||
| BC115807.1 | - | 3 | 336 | 40106016 | ||

1810022K09Rik
Genomic Map