siRNA Details
| Gene Symbol: Cd209f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026956.2 | NP_081232.2 | 6, 7 | 545 | ||
| GenBank mRNA | AK007638.1 | - | 6, 7 | 602 | ||
| BC021323.1 | - | 6, 7 | 627 | |||
| BC046292.1 | - | 4, 5 | 586 | |||
| BC052782.1 | - | 6, 7 | 536 | |||
| BC099497.1 | - | 6, 7 | 540 | |||

Cd209f
Genomic Map