siRNA Details
Gene Symbol: 2010016I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040034.1 | - | 1 | 1378 | ||
AK079963.1 | - | 2 | 768 | |||
AK080971.1 | - | 2 | 1468 | |||
AK140363.1 | - | 2 | 1468 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2010016I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040034.1 | - | 1 | 1378 | ||
AK079963.1 | - | 2 | 768 | |||
AK080971.1 | - | 2 | 1468 | |||
AK140363.1 | - | 2 | 1468 |