siRNA Details
| Gene Symbol: Zfp746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163475.1 | NP_001156947.1 | 7 | 3606 | ||
| GenBank mRNA | AK013039.1 | - | 1 | 597 | ||
| AK156312.1 | - | 7 | 3607 | |||
| BC065410.1 | - | 1 | 755 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp746| Gene Symbol: Zfp746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163475.1 | NP_001156947.1 | 7 | 3606 | ||
| GenBank mRNA | AK013039.1 | - | 1 | 597 | ||
| AK156312.1 | - | 7 | 3607 | |||
| BC065410.1 | - | 1 | 755 | |||