siRNA Details
| Gene Symbol: Rhobtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081347.1 | NP_001074816.1 | 11 | 4033 | ||
| NM_001252636.1 | NP_001239565.1 | 11 | 4016 | |||
| NM_001252637.1 | NP_001239566.1 | 11 | 4137 | |||
| NM_001252638.1 | NP_001239567.1 | 12 | 4196 | |||
| GenBank mRNA | AK014194.1 | - | 1 | 1359 | ||

Rhobtb1
Genomic Map