siRNA Details
| Gene Symbol: 1700001L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027035.1 | NP_081311.1 | 3 | 1395 | ||
| GenBank mRNA | AK138387.1 | - | 3 | 1396 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001L19Rik| Gene Symbol: 1700001L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027035.1 | NP_081311.1 | 3 | 1395 | ||
| GenBank mRNA | AK138387.1 | - | 3 | 1396 | ||