siRNA Details
Gene Symbol: 1700007K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027040.1 | NP_081316.1 | 3 | 382 | ||
GenBank mRNA | AK005731.1 | - | 3 | 392 | ||
AY927869.1 | - | 3 | 327 | |||
BC048581.1 | - | 3 | 363 | |||
BC099566.1 | - | 3 | 382 | 30916995 |
