siRNA Details
| Gene Symbol: 1700008O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027049.1 | NP_081325.1 | 4 | 385 | ||
| GenBank mRNA | AK005784.1 | - | 4 | 386 | ||
| BC127243.1 | - | 2 | 501 | |||
| BC127244.1 | - | 2 | 501 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700008O03Rik| Gene Symbol: 1700008O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027049.1 | NP_081325.1 | 4 | 385 | ||
| GenBank mRNA | AK005784.1 | - | 4 | 386 | ||
| BC127243.1 | - | 2 | 501 | |||
| BC127244.1 | - | 2 | 501 | |||