siRNA Details
| Gene Symbol: 1700012P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027056.1 | NP_081332.1 | 1 | 48 | ||
| GenBank mRNA | AK005933.1 | - | 1 | 49 | ||
| BC099569.1 | - | 1 | 48 | 30918007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700012P22Rik| Gene Symbol: 1700012P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027056.1 | NP_081332.1 | 1 | 48 | ||
| GenBank mRNA | AK005933.1 | - | 1 | 49 | ||
| BC099569.1 | - | 1 | 48 | 30918007 | ||