siRNA Details
| Gene Symbol: 1700017D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027058.1 | NP_081334.1 | 4 | 669 | ||
| GenBank mRNA | AK006056.1 | - | 4 | 675 | ||
| BU938748.1 | - | 4 | 672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700017D01Rik| Gene Symbol: 1700017D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027058.1 | NP_081334.1 | 4 | 669 | ||
| GenBank mRNA | AK006056.1 | - | 4 | 675 | ||
| BU938748.1 | - | 4 | 672 | |||