siRNA Details
| Gene Symbol: H2bfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027067.2 | NP_081343.1 | 1 | 703 | ||
| GenBank mRNA | AK005982.1 | - | 1 | 704 | ||
| BC117076.1 | - | 1 | 694 | 40126395 | ||
| BC117080.1 | - | 1 | 694 | 40126399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2bfm| Gene Symbol: H2bfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027067.2 | NP_081343.1 | 1 | 703 | ||
| GenBank mRNA | AK005982.1 | - | 1 | 704 | ||
| BC117076.1 | - | 1 | 694 | 40126395 | ||
| BC117080.1 | - | 1 | 694 | 40126399 | ||