siRNA Details
| Gene Symbol: Gja5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019280.1 | NP_062153.1 | 2 | 245 | ||
| XM_008761328.1 | XP_008759550.1 | 2 | 3384 | |||
| GenBank mRNA | AF021806.1 | - | 2 | 245 | ||
| BC070935.1 | - | 2 | 267 | 7097997 | ||
| M83092.1 | - | 1 | 155 | |||

Gja5
Genomic Map