siRNA Details
| Gene Symbol: Krtap3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023511.1 | NP_076000.1 | 1 | 571 | ||
| GenBank mRNA | AK009119.1 | - | 1 | 320 | ||
| BC060278.1 | - | 1 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap3-1| Gene Symbol: Krtap3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023511.1 | NP_076000.1 | 1 | 571 | ||
| GenBank mRNA | AK009119.1 | - | 1 | 320 | ||
| BC060278.1 | - | 1 | 574 | |||