siRNA Details
| Gene Symbol: Ttc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033149.2 | NP_001028321.1 | 3 | 1995 | ||
| GenBank mRNA | AK133456.1 | - | 3 | 1996 | ||
| AK161499.1 | - | 3 | 1795 | |||
| AK172918.1 | - | 2087 | ||||
| BC028891.1 | - | 4 | 2051 | |||

Ttc9
Genomic Map