siRNA Details
Gene Symbol: Trnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081156.2 | NP_001074625.1 | 2 | 1123 | ||
GenBank mRNA | AY142705.1 | - | 2 | 537 | ||
BC049168.1 | - | 2 | 1079 | 6491759 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081156.2 | NP_001074625.1 | 2 | 1123 | ||
GenBank mRNA | AY142705.1 | - | 2 | 537 | ||
BC049168.1 | - | 2 | 1079 | 6491759 |