siRNA Details
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081058.2 | NP_001074527.1 | 14 | 6449 | ||
NM_027118.1 | NP_081394.1 | 14 | 6269 | |||
GenBank mRNA | AK078640.1 | - | 1 | 1290 | ||
AK144792.1 | - | 1 | 284 | |||
AK148986.1 | - | 1 | 2070 |
