siRNA Details
| Gene Symbol: Plekhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033150.1 | NP_001028322.1 | 8 | 957 | ||
| GenBank mRNA | AK122386.1 | - | 981 | |||
| AK156652.1 | - | 5 | 579 | |||
| BC058984.1 | - | 6 | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhm2| Gene Symbol: Plekhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033150.1 | NP_001028322.1 | 8 | 957 | ||
| GenBank mRNA | AK122386.1 | - | 981 | |||
| AK156652.1 | - | 5 | 579 | |||
| BC058984.1 | - | 6 | 548 | |||