siRNA Details
| Gene Symbol: Fam89a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081120.1 | NP_001074589.1 | 2 | 940 | ||
| GenBank mRNA | AK009550.1 | - | 1 | 514 | ||
| BC023460.1 | - | 2 | 781 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam89a| Gene Symbol: Fam89a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081120.1 | NP_001074589.1 | 2 | 940 | ||
| GenBank mRNA | AK009550.1 | - | 1 | 514 | ||
| BC023460.1 | - | 2 | 781 | |||