siRNA Details
| Gene Symbol: Cd164l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027152.1 | NP_081428.1 | 3 | 445 | ||
| GenBank mRNA | AK009888.1 | - | 3 | 371 | ||
| AK139809.1 | - | 3 | 446 | |||
| BC111824.1 | - | 3 | 368 | 40055637 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd164l2| Gene Symbol: Cd164l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027152.1 | NP_081428.1 | 3 | 445 | ||
| GenBank mRNA | AK009888.1 | - | 3 | 371 | ||
| AK139809.1 | - | 3 | 446 | |||
| BC111824.1 | - | 3 | 368 | 40055637 | ||