siRNA Details
| Gene Symbol: Cd164l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027152.1 | NP_081428.1 | 1 | 221 | ||
| GenBank mRNA | AK009888.1 | - | 1 | 147 | ||
| AK139809.1 | - | 1 | 222 | |||
| BC111824.1 | - | 1 | 144 | 40055637 | ||
| CJ072306.1 | - | 1 | 224 | |||

Cd164l2
Genomic Map