siRNA Details
| Gene Symbol: Tmem52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253856.1 | NP_001240785.1 | 5 | 536 | ||
| NM_001253858.1 | NP_001240787.1 | 4 | 494 | |||
| NM_027161.2 | NP_081437.1 | 5 | 554 | |||
| GenBank mRNA | AK009779.1 | - | 5 | 517 | ||
| BC144992.1 | - | 5 | 553 | 9053540 | ||
| BC150685.1 | - | 5 | 534 | 9087596 | ||

Tmem52
Genomic Map