siRNA Details
| Gene Symbol: Trip13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027182.2 | NP_081458.1 | 11 | 1307 | ||
| GenBank mRNA | AK010336.1 | - | 11 | 1269 | ||
| AK142463.1 | - | 1280 | ||||
| AK146877.1 | - | 11 | 1308 | |||
| AK151568.1 | - | 11 | 1243 | |||
| AK212754.1 | - | 3 | 345 | |||
| BC023834.1 | - | 5 | 515 | |||
| BC064315.1 | - | 11 | 1230 | |||
| BC126946.1 | - | 11 | 1206 | 40090292 | ||

Trip13
Genomic Map