siRNA Details
| Gene Symbol: Zswim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027198.1 | NP_081474.1 | 4 | 336 | ||
| GenBank mRNA | AK010472.1 | - | 4 | 316 | ||
| BC109343.2 | - | 4 | 294 | 40053371 | ||
| BQ554850.1 | - | 4 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim7| Gene Symbol: Zswim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027198.1 | NP_081474.1 | 4 | 336 | ||
| GenBank mRNA | AK010472.1 | - | 4 | 316 | ||
| BC109343.2 | - | 4 | 294 | 40053371 | ||
| BQ554850.1 | - | 4 | 339 | |||