siRNA Details
| Gene Symbol: 1810046K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027217.1 | NP_081493.1 | 5 | 1305 | ||
| GenBank mRNA | EH105864.1 | - | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810046K07Rik| Gene Symbol: 1810046K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027217.1 | NP_081493.1 | 5 | 1305 | ||
| GenBank mRNA | EH105864.1 | - | 214 | |||