siRNA Details
| Gene Symbol: Ndufa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027244.1 | NP_081520.1 | 4 | 2100 | ||
| GenBank mRNA | BC053075.1 | - | 3 | 2581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufa11| Gene Symbol: Ndufa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027244.1 | NP_081520.1 | 4 | 2100 | ||
| GenBank mRNA | BC053075.1 | - | 3 | 2581 | ||