siRNA Details
| Gene Symbol: Igsf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027308.2 | NP_081584.1 | 5 | 1659 | ||
| GenBank mRNA | AK033677.1 | - | 5 | 1660 | ||
| AK088927.1 | - | 6 | 1615 | |||
| AK135657.1 | - | 1465 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igsf23| Gene Symbol: Igsf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027308.2 | NP_081584.1 | 5 | 1659 | ||
| GenBank mRNA | AK033677.1 | - | 5 | 1660 | ||
| AK088927.1 | - | 6 | 1615 | |||
| AK135657.1 | - | 1465 | ||||