siRNA Details
| Gene Symbol: Lrch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081255.1 | NP_001074724.1 | 21 | 4025 | ||
| GenBank mRNA | AK134026.1 | - | 11 | 2656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrch3| Gene Symbol: Lrch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081255.1 | NP_001074724.1 | 21 | 4025 | ||
| GenBank mRNA | AK134026.1 | - | 11 | 2656 | ||