siRNA Details
Gene Symbol: 2210016F16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027335.1 | NP_081611.1 | 4 | 2777 | ||
GenBank mRNA | AK052253.1 | - | 4 | 2780 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2210016F16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027335.1 | NP_081611.1 | 4 | 2777 | ||
GenBank mRNA | AK052253.1 | - | 4 | 2780 |