siRNA Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081079.1 | NP_001074548.1 | 7 | 2436 | ||
| GenBank mRNA | AK034467.1 | - | 1 | 1068 | ||
| AK034496.1 | - | 1 | 1139 | |||
| AK037156.1 | - | 1 | 1321 | |||
| AK078578.1 | - | 1 | 1019 | |||
| AK142127.1 | - | 1 | 1186 | |||

Ogfrl1
Genomic Map