siRNA Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081079.1 | NP_001074548.1 | 7 | 3091 | ||
| GenBank mRNA | AK034467.1 | - | 1 | 1723 | ||
| AK034496.1 | - | 1 | 1794 | |||
| AK037156.1 | - | 1 | 1977 | |||
| AK142127.1 | - | 1 | 1841 | |||

Ogfrl1
Genomic Map