siRNA Details
| Gene Symbol: Cd209g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027343.3 | NP_081619.3 | 7, 8 | 641 | ||
| GenBank mRNA | AK010071.1 | - | 6, 7 | 602 | ||
| AM286437.1 | - | 7, 8 | 644 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd209g| Gene Symbol: Cd209g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027343.3 | NP_081619.3 | 7, 8 | 641 | ||
| GenBank mRNA | AK010071.1 | - | 6, 7 | 602 | ||
| AM286437.1 | - | 7, 8 | 644 | |||