siRNA Details
Gene Symbol: Mkrn2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101431.1 | NP_001094901.1 | 4 | 1862 | ||
GenBank mRNA | AK028760.1 | - | 3 | 1735 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mkrn2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101431.1 | NP_001094901.1 | 4 | 1862 | ||
GenBank mRNA | AK028760.1 | - | 3 | 1735 |