siRNA Details
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027376.3 | NP_081652.2 | 2 | 356 | ||
| GenBank mRNA | AK011195.1 | - | 2 | 305 | ||
| AK033247.1 | - | 2 | 343 | |||
| AK045745.1 | - | 351 | ||||
| AK134650.1 | - | 2 | 356 | |||
| AK168939.1 | - | 2 | 351 | |||
| BC045601.1 | - | 2 | 342 | |||

Fuz
Genomic Map