siRNA Details
| Gene Symbol: Arl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197995.2 | NP_932112.2 | 5 | 1635 | ||
| GenBank mRNA | AK140077.1 | - | 3 | 1801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl16| Gene Symbol: Arl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197995.2 | NP_932112.2 | 5 | 1635 | ||
| GenBank mRNA | AK140077.1 | - | 3 | 1801 | ||