siRNA Details
| Gene Symbol: Tecpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027410.1 | NP_081686.1 | 10 | 1762 | ||
| GenBank mRNA | AB093295.1 | - | 9 | 1931 | ||
| AK032858.1 | - | 10 | 1736 | |||
| AK034020.1 | - | 9 | 2341 | |||
| AK169888.1 | - | 10 | 1709 | |||
| AK170108.1 | - | 10 | 1709 | |||
| AK195750.1 | - | 1 | 17 | |||
| AK210140.1 | - | 1 | 17 | |||
| AK218703.1 | - | 1 | 125 | |||
| BC046968.1 | - | 10 | 1762 | 6306206 | ||
| BC052185.1 | - | 4 | 2339 | |||

Tecpr1
Genomic Map